Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone
TRC 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone
TRC 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone
Catalog # 77989-198
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone
Catalog # 77989-198
Supplier:
LGC Standards
Supplier Number:
TRC-M325750-10MG
New Product
CAS Number:
Specifications
CAS No.:
64091-91-4
Description:
4-(Methylnitrosamino)-1-(3-pyridyl)-1-butanone
Size:
10 mg
Specifications
About this item
Product Details
Product Details