Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Tetramethyl Bisphenol A
TRC Tetramethyl Bisphenol A
77994-135
:
LGC Standards
New Product
:
TRC Tetramethyl Bisphenol A
77994-135
:
LGC Standards
:
TRC-T300930-1G
New Product
:
CAS No.:
5613-46-7
Description:
Tetramethyl Bisphenol A
Size:
1 g