Order Entry
Canada
ContactUsLinkComponent
Dr. Ehrenstorfer Microplastics Polypropylene PP nominal 35 μm) (Dv50 = 120 μm)
Dr. Ehrenstorfer Microplastics Polypropylene PP nominal 35 μm) (Dv50 = 120 μm)
Catalog # 77976-080
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Dr. Ehrenstorfer Microplastics Polypropylene PP nominal 35 μm) (Dv50 = 120 μm)
Catalog # 77976-080
Supplier:  LGC Standards
Supplier Number:  DRE-MP-PP-35
New Product
CAS Number:  

Specifications

  • CAS No.:
    9003-07-0
  • Description:
    Microplastics polypropylene PP nominal 35 μm (Dv50 = 120 μm; Dn50 = 3 μm)
  • Size:
    50 mg
  • Shipping conditions:
    20 °C
  • Formula:
    n(C₃H₆)
  • Formula weight:
    42,08 g/mol
  • Storage temperature:
    20 °C
  • Certification:
    ISO/IEC 17025

Specifications

About this item