ContactUsLinkComponent
Paragon Scientific 1413 µS/cm @ 25 °C, Conductivity (NIST Traceable, ACS)
  77975-552
New Product
 :  
Paragon Scientific 1413 µS/cm @ 25 °C, Conductivity (NIST Traceable, ACS)
  77975-552
 :  ALK-COND1413-25-500ML
New Product
 :  
  • Description:
    Conductivity standard CRM
  • Size:
    500 ml
  • Reference Temperature:
    25 °C
  • Certification:
    ISO 17034
  • Conductivity:
    1413 µS/cm