Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Phenobarbital-d5 (100 μg/ml in Methanol)
TRC Phenobarbital-d5 (100 μg/ml in Methanol)
TRC Phenobarbital-d5 (100 μg/ml in Methanol)
77987-713
:
LGC Standards
New Product
:
TRC Phenobarbital-d5 (100 μg/ml in Methanol)
77987-713
:
LGC Standards
:
TRC-KIT0222-5X1ML
New Product
:
Matrix:
Methanol
CAS No.:
73738-05-3
Description:
Phenobarbital-d5 (100 μg/ml in Methanol)
Size:
5×1 ml