Orders LinkContactUsLinkComponent
Filming Amines Acidifier Solution Pack
Filming Amines Acidifier Solution Pack
Rose Bengal
Catalog # 77674-530
Supplier:   CHEMetrics
Supplier #:  A-1010
Filming Amines Acidifier Solution Pack
Catalog # 77674-530
Supplier:  CHEMetrics
CAS Number:  
Filming Amines Acidifier Solution Pack
Filming Amines Acidifier Solution Pack
Rose Bengal
CAS Number:  
Price
$48.63
Log in for availability
Pack of 6 (20ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A-1010 Filming Amines Acidifier Solution Pack contains six 20 ml bottles of pH adjustment chemical solution. This is enough acidifier solution to perform at least 200 tests. This water pH adjuster pack is compatible with K-1006 and K-1013 Filming Amine Test Kits.
Specifications
Specifications
  • Chemical method:
    Rose Bengal
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)