Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Simvastatin-d6
TRC Simvastatin-d6
TRC Simvastatin-d6
Catalog # 77993-175
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Simvastatin-d6
Catalog # 77993-175
Supplier:
LGC Standards
Supplier Number:
TRC-S485002-2.5MG
New Product
CAS Number:
Specifications
CAS No.:
1002347-71-8
Description:
Simvastatin-d6
Size:
2.5 mg
Specifications
About this item
Product Details
Product Details