TRC Pyruvic Acid
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:127-17-3
Specifications
Frequently Bought Together
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Acetone ≥99.5% (by GC, corrected for water content), GR ACS, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
tri-Sodium citrate dihydrate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Ozone AccuVac® Ampules, MR, Hach
Supplier: Hach
XLD ( Xylose Lysine Deoxycholate) Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Hydrazinium sulfate ≥99.0% ACS
Supplier: Thermo Scientific Chemicals
Exel International® Hypodermic Needles, Air-Tite Products
Supplier: Air-Tite
SYRINGE WITH NEEDLE 5ML
Supplier: AMX INDUSTRIAL TRADE MX
Human Recombinant CNTF (from <i>E. coli</i>)
Supplier: Peprotech
LabKlenz™ 250 Acid Detergents
Supplier: Steris
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences
Reagent Sets for Colorimeters, Hach
Supplier: Hach
SYRINGE XS 5ML SALINE PK30
Supplier: BD