TRC Benzalkonium Chloride
New Product
- CAS No.:63449-41-2
Frequently Bought Together
Ethyl Alcohol Denatured
: Ward's Science
SYRINGE FOR INSULIN WITH NEEDLE 1ML
: AMX INDUSTRIAL TRADE MX
Electrode Cleaning and Storage Solutions, Oakton®, Cole Parmer
: Antyila Scientific
96-Well and 384-Well Color Orienters, Electron Microscopy Sciences
: Electron Microscopy Sciences
HyClone™ SFM4CHO media, Cytiva
: Cytiva
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
: VWR International
Toluene ACS
: INNOVA CHEMICALS CA
Acetic acid glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
: AVANTOR PERFORMANCE MATERIAL LLC
pH Indicator Papers, Duotest, Machery Nagel
: Macherey-Nagel