Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC (3R)-2,3-Dihydro-3-(2-propyn-1-ylamino)-1H-inden-1-one
TRC (3R)-2,3-Dihydro-3-(2-propyn-1-ylamino)-1H-inden-1-one
TRC (3R)-2,3-Dihydro-3-(2-propyn-1-ylamino)-1H-inden-1-one
Catalog # 77982-790
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC (3R)-2,3-Dihydro-3-(2-propyn-1-ylamino)-1H-inden-1-one
Catalog # 77982-790
Supplier:
LGC Standards
Supplier Number:
TRC-D449765-5MG
New Product
CAS Number:
Specifications
CAS No.:
1312077-05-6
Description:
(3R)-2,3-Dihydro-3-(2-propyn-1-ylamino)-1H-inden-1-one
Size:
5 mg
Specifications
About this item
Product Details
Product Details