Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Ritodrine Hydrochloride
TRC Ritodrine Hydrochloride
TRC Ritodrine Hydrochloride
77992-833
:
LGC Standards
New Product
:
TRC Ritodrine Hydrochloride
77992-833
:
LGC Standards
:
TRC-R533700-250MG
New Product
:
CAS No.:
23239-51-2
Description:
Ritodrine Hydrochloride
Size:
250 mg