TRC 5-(1-Methyl-4-piperidinyl)-5H-dibenzo[a,d]cyclohepten-5-ol
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:3967-32-6
Specifications
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Liquinox® Critical Cleaning Liquid Detergents
Supplier: Alconox
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
Sodium sulfate, anhydrous ≥99.0%, white powder ACS, VWR Chemicals BDH®
Supplier: VWR International
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Supplier: Corning
Difco™ Tryptic Soy Agar, BD Biosciences
Supplier: BD
VWR® Premium Frozen Section Compound
Supplier: VWR International
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: HARDY DIAGNOSTICS CA
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Q5® High-Fidelity DNA Polymerase
Supplier: New England Biolabs (NEB)
Sol-M® Standard Insulin Syringes
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Pierce SnakeSkin™ Pleated Dialysis Tubing, MWCO 3500 to 10000, Thermo Scientific
Supplier: Thermo Scientific