Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 10,11-Epoxide Carbamazepine- d10
TRC 10,11-Epoxide Carbamazepine- d10
TRC 10,11-Epoxide Carbamazepine- d10
Catalog # 77979-417
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 10,11-Epoxide Carbamazepine- d10
Catalog # 77979-417
Supplier:
LGC Standards
Supplier Number:
TRC-C175856-1MG
New Product
CAS Number:
Specifications
CAS No.:
1219804-16-6
Description:
10,11-Epoxide Carbamazepine- d10
Size:
1 mg
Specifications
About this item
Product Details
Product Details