Paragon Scientific Sulfur @ 0.3% (3000 ppm) in light mineral oil
Catalog # 77975-879
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Light Mineral Oil
- CAS No.:7704-34-9
- Description:Sulfur @ 0.3% (3000 ppm) in light mineral oil
- Size:100 ml
- Certification:ISO 17034
- Storage conditions:RT
- Shipping conditions:RT
- Molecular weight:32.065