Dr. Ehrenstorfer Perfluorooctanoic acid 13C8 50 µg/ml in Methanol:Water
Catalog # 77976-048
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol:Water
- CAS No.:1350614-84-4
- Description:Perfluorooctanoic acid 13C8 50 µg/ml in Methanol:Water
- Size:1 ml
- Shipping conditions:20 °C
- Formula:¹³C₈HF₁₅O₂
- Formula weight:422,01 g/mol
- Storage temperature:−18 °C
- Certification:ISO 17034
- Certification:ISO 17034