TRC Bisphenol F-d10
Fournisseur: LGC Standards
New Product
Certificats
À propos de cet article
Spécifications
- CAS No.:1794786-93-8
Spécifications
Frequently Bought Together
Puritan® Standard Polyester Tipped Applicator, Polystyrene Handle, Sterile, Puritan Medical Products
Fournisseur: Puritan Medical Products
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Fournisseur: Thermo Scientific
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Fournisseur: Air-Tite
PerfeCTa® SYBR® Green FastMix® Reaction Mixes, Quantabio
Fournisseur: Quantabio
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Fournisseur: Air-Tite
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
BD Syringes only with Luer-Lok™, Slip or Eccentric tips
Fournisseur: BD
Hydrochloric acid 36,5 - 38,0% ACS
Fournisseur: Anachemia
Exel International® Hypodermic Needles, Air-Tite Products
Fournisseur: Air-Tite
Human Recombinant CNTF (from <i>E. coli</i>)
Fournisseur: Peprotech
Monarch® RNA Cleanup Kits (50 μg), New England Biolabs
Fournisseur: New England Biolabs (NEB)
Accessories for Gas Regulators
Fournisseur: VWR International
TapeStation Systems Parts and Accessories
Fournisseur: AGILENT TECHNOLOGIES (GENOMICS) CA
Acid, Caustic and Solvent Spill Kit
Fournisseur: Ward's Science
Sol-M® Hypodermic Needles
Fournisseur: SOL-MILLENNIUM CANADA, INC. CA
BD Vacutainer® Blood Collection Needles, BD Diagnostics
Fournisseur: BD
E.Z.N.A.® Total RNA Kit I, Omega Bio-tek®
Fournisseur: Omega Bio-Tek
Saline, 0.85%
Fournisseur: Hardy Diagnostics