Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 5-Carboxy-N-phenyl-2-1H-pyridone
TRC 5-Carboxy-N-phenyl-2-1H-pyridone
TRC 5-Carboxy-N-phenyl-2-1H-pyridone
Catalog # 77979-485
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 5-Carboxy-N-phenyl-2-1H-pyridone
Catalog # 77979-485
Supplier:
LGC Standards
Supplier Number:
TRC-C179020-50MG
New Product
CAS Number:
Specifications
CAS No.:
77837-08-2
Description:
5-Carboxy-N-phenyl-2-1H-pyridone
Size:
50 mg
Specifications
About this item
Product Details
Product Details