Paragon Scientific Sulfur @ 3% (30000 ppm) in heavy mineral oil
77975-770
New Product
:
- Matrix:Heavy Mineral Oil
- CAS No.:7704-34-9
- Description:Sulfur @ 3% (30000 ppm) in heavy mineral oil
- Size:100 ml
- Certification:ISO 17034
- Storage conditions:RT
- Shipping conditions:RT
- Molecular weight:32.065