Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC L-Thyronine, 95%
TRC L-Thyronine, 95%
TRC L-Thyronine, 95%
Catalog # 77995-008
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC L-Thyronine, 95%
Catalog # 77995-008
Supplier:
LGC Standards
Supplier Number:
TRC-T922700-50MG
New Product
CAS Number:
Specifications
CAS No.:
1596-67-4
Description:
L-Thyronine, 95%
Size:
50 mg
Specifications
About this item
Product Details
Product Details