TRC Perfluorohexanesulfonamide (~75%)
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:41997-13-1
Specifications
Frequently Bought Together
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Leakbuster™ Specimen Containers
Supplier: Starplex
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Acetone ACS for production, VWR Chemicals BDH®
Supplier: VWR International
Masterflex® Union Fittings, Hose Barb, Tee, Polypropylene, Avantor®
Supplier: VWR International
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.9%, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Hexane (mixture of isomers) ≥98.5%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
Calcium chloride, dried 97%, powder
Supplier: Thermo Scientific Chemicals
GASTIGHT® 1000 Series Syringes, Hamilton
Supplier: Hamilton
Accessories for Blood Bank Plasma Thawer, Boekel Scientific
Supplier: Boekel
Potassium hexafluorophosphate ≥99%
Supplier: Thermo Scientific Chemicals
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Feather™ Scalpel Handle with Blade, Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Conflikt® Ready-to-use Disinfectant
Supplier: Decon Labs