Paragon Scientific Sulfur @ 0.1% (1000 ppm) in odorless kerosene
Catalog # 77975-837
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Odourless Kerosene
- CAS No.:7704-34-9
- Description:Sulfur @ 0.1% (1000 ppm) in odorless kerosene
- Size:100 ml
- Certification:ISO 17034
- Storage conditions:RT
- Shipping conditions:RT
- Molecular weight:32.065