Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Tetraglyme-d6
TRC Tetraglyme-d6
TRC Tetraglyme-d6
Catalog # 77994-062
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Tetraglyme-d6
Catalog # 77994-062
Supplier:
LGC Standards
Supplier Number:
TRC-T292997-10MG
New Product
CAS Number:
Specifications
CAS No.:
1216628-15-7
Description:
Tetraglyme-d6
Size:
10 mg
Specifications
About this item
Product Details
Product Details