TRC Fipronil
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:120068-37-3
Specifications
Frequently Bought Together
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
VWR® Disposable Petri Dishes
Supplier: VWR International
Single-Use Needles, BD Medical
Supplier: BD
Corning® Cell and Tissue Adhesive, Corning
Supplier: Corning
Corning® DMEM/Ham's F-12 50/50 Mix Products, Corning
Supplier: Corning
Diphenylacetonitrile ≥99.0%
Supplier: TCI America
Corning® Trypsin
Supplier: Corning
Reagent Sets for Colorimeters, Hach
Supplier: Hach
Formaldehyde (Paraformaldehyde) 16% in aqueous solution, EM Grade
Supplier: Electron Microscopy Science
m Endo LES Agar, for Membrane Filtration, Hardy Diagnostics
Supplier: Hardy Diagnostics