Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 6-(tert-Butylamino)-1,3,5-triazine-2,4(1H,3H)-dione
TRC 6-(tert-Butylamino)-1,3,5-triazine-2,4(1H,3H)-dione
TRC 6-(tert-Butylamino)-1,3,5-triazine-2,4(1H,3H)-dione
77978-201
:
LGC Standards
New Product
:
TRC 6-(tert-Butylamino)-1,3,5-triazine-2,4(1H,3H)-dione
77978-201
:
LGC Standards
:
TRC-B185955-10MG
New Product
:
CAS No.:
309923-18-0
Description:
6-(tert-Butylamino)-1,3,5-triazine-2,4(1H,3H)-dione
Size:
10 mg