Foalwatch Accessory Solution
Product Details & Documents
QTY: 1. 1 bottle of accessory solution for the FoalWatch test kit.
Specifications
Specifications
- Chemical method:EGTA (calcium)
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)