CHEMets® Silica Refill
Catalog # 77674-908
Supplier: CHEMetrics
New Product
CAS Number:
Specifications
- Chemical method:Heteropoly blue
- Application:Boiler water;Cooling water;Natural water;Waste water;High purity water
Specifications
About this item
This product is a refill for K-9010 Silica CHEMets visual kit for the analysis of silica in water. It contains 30 self-filling CHEMets ampoules packaged securely in cardboard trays and box. R-9010 Silica CHEMets refill is compatible with C-9001 Low Range Silica Comparator and C-9010 high range Silica Comparator.
- Method: Heteropoly blue