$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Standards
Organic Standards
TRC N,N-Di(2-hydroxypropyl)-p-toluidine (>85%)
TRC N,N-Di(2-hydroxypropyl)-p-toluidine (>85%)
77982-677
:
LGC Standards
New Product
:
TRC N,N-Di(2-hydroxypropyl)-p-toluidine (>85%)
77982-677
:
LGC Standards
:
TRC-D446665-10G
New Product
:
CAS No.:
38668-48-3
Description:
N,N-Di(2-hydroxypropyl)-p-toluidine (>85%)
Size:
10 g