TRC N-gamma-Acetyl-N-2-formyl-5-methoxykynurenamine
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:52450-38-1
Specifications
Frequently Bought Together
Calcium chloride, anhydrous ≥96% ACS
Supplier: VWR
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Razor Blades
Supplier: VWR International
Water, HiPerSolv CHROMANORM for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Phenolphthalein 1% (w/v) in 95% (v/v) alcohol indicator, neutralized
Supplier: Ricca Chemical
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Buffer, TBS Solution 10X Concentrate (TRIS Buffered Saline), G-Biosciences
Supplier: G-Biosciences
LabKlenz™ 100 Alkaline Detergents
Supplier: Steris
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
HistoGel™ Specimen Processing Gel, Epredia
Supplier: Thermo Scientific
Acetaldehyde 99%
Supplier: Thermo Scientific Chemicals
E.Z.N.A.® Plasmid Mini Kit I, Omega Bio-tek®
Supplier: Omega Bio-Tek
Endothelial Cell Growth Media Kits, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
Dithiothreitol (DTT, Cleland's reagent), SPUTOLYSIN®
Supplier: MilliporeSigma
Amino Acid F Reagent Powder Pillows, 10 ml, Hach
Supplier: Hach
Sodium hydroxide, pellets NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC