Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 4-(p-Chloro-alpha-phenylbenzyl)-1-piperazineethanol
TRC 4-(p-Chloro-alpha-phenylbenzyl)-1-piperazineethanol
TRC 4-(p-Chloro-alpha-phenylbenzyl)-1-piperazineethanol
Catalog # 77980-041
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 4-(p-Chloro-alpha-phenylbenzyl)-1-piperazineethanol
Catalog # 77980-041
Supplier:
LGC Standards
Supplier Number:
TRC-C377565-250MG
New Product
CAS Number:
Specifications
CAS No.:
109806-71-5
Description:
4-(p-Chloro-alpha-phenylbenzyl)-1-piperazineethanol
Size:
250 mg
Specifications
About this item
Product Details
Product Details