Order Entry
Canada
Orders LinkContactUsLinkComponent
CHEMets® Nitrite Refill

CHEMets® Nitrite Refill

Azo dye formation
Catalog # 77674-902
Supplier:   CHEMetrics
Supplier #:  R-7006
CHEMets® Nitrite Refill
Catalog # 77674-902
Supplier:  CHEMetrics
CAS Number:  
CHEMets® Nitrite Refill
CHEMets® Nitrite Refill

Azo dye formation

CAS Number:  
Price
$64.06
Log in for availability
Pack of 30
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

This product is a refill for K-7006 Nitrite CHEMets visual kit for the analysis of nitrite in water. It contains 30 self-filling CHEMets ampoules packaged securely in cardboard trays and box. R-7006 Nitrite CHEMets refill is compatible with C-7006 Nitrite low range comparator and C-7007 Nitrite high range comparator.

  • Method: Azo Dye Formation (NED)
Specifications

Specifications

  • Chemical method:
    Azo dye formation
  • Application:
    Industrial waste water influent and effluent;Industrial process water;Boiler water;Cooling water;Surface and ground water

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.