Dr. Ehrenstorfer Microplastics Polyethylene PE nominal 65 μm) (Dv50 = 130 μm)
77976-076
New Product
:
- CAS No.:9002-88-4
- Description:Microplastics polyethylene PE nominal 65 μm (Dv50 = 130 μm; Dn50 = 3 μm)
- Size:50 mg
- Shipping conditions:20 °C
- Formula:C₂H₄
- Formula weight:28,053 g/mol
- Storage temperature:20 °C
- Certification:ISO/IEC 17025
- Certification:ISO/IEC 17025