Paragon Scientific Sulfur @ 0.25% (2500 ppm) in heavy mineral oil
Catalog # 77975-764
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Heavy Mineral Oil
- CAS No.:7704-34-9
- Description:Sulfur @ 0.25% (2500 ppm) in heavy mineral oil
- Size:100 ml
- Certification:ISO 17034
- Storage conditions:RT
- Shipping conditions:RT
- Molecular weight:32.065