Paragon Scientific ASTM D7039, Sulfur set A in odorless kerosene: S @ blank, 0.0005, 0.005, 0.025, 0.05%
Catalog # 77975-616
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Odourless Kerosene
- CAS No.:7704-34-9
- Description:ASTM D7039, Sulfur set A in odorless kerosene: S @ blank, 0.0005, 0.005, 0.025, 0.05%
- Size:5×100 ml
- Certification:ISO 17034
- Storage conditions:RT
- Shipping conditions:RT
- Molecular weight:32.065