Order Entry
Canada
ContactUsLinkComponent
Paragon Scientific Sulfur @ 1.5% (15000 ppm) in heavy mineral oil
Paragon Scientific Sulfur @ 1.5% (15000 ppm) in heavy mineral oil
Catalog # 77975-768
Supplier:  LGC Standards
New Product
CAS Number:  
Paragon Scientific Sulfur @ 1.5% (15000 ppm) in heavy mineral oil
Catalog # 77975-768
Supplier:  LGC Standards
Supplier Number:  ALK-HMSU-16
New Product
CAS Number:  

Specifications

  • Matrix:
    Heavy Mineral Oil
  • CAS No.:
    7704-34-9
  • Description:
    Sulfur @ 1.5% (15000 ppm) in heavy mineral oil
  • Size:
    100 ml
  • Certification:
    ISO 17034
  • Storage conditions:
    RT
  • Shipping conditions:
    RT
  • Molecular weight:
    32.065

Specifications

About this item