Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Monopropylheptylphthalate 6-Hydroxy-d4
TRC Monopropylheptylphthalate 6-Hydroxy-d4
TRC Monopropylheptylphthalate 6-Hydroxy-d4
77989-733
:
LGC Standards
New Product
:
TRC Monopropylheptylphthalate 6-Hydroxy-d4
77989-733
:
LGC Standards
:
TRC-M567077-1MG
New Product
:
CAS No.:
1412411-11-0
Description:
Monopropylheptylphthalate 6-Hydroxy-d4
Size:
1 mg