Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Nandrolone-d3 (1.0 mg/ml in Acetonitrile)
TRC Nandrolone-d3 (1.0 mg/ml in Acetonitrile)
TRC Nandrolone-d3 (1.0 mg/ml in Acetonitrile)
77987-751
:
LGC Standards
New Product
:
TRC Nandrolone-d3 (1.0 mg/ml in Acetonitrile)
77987-751
:
LGC Standards
:
TRC-KIT6470-1X1ML
New Product
:
Matrix:
Acetonitrile
CAS No.:
120813-22-1
Description:
Nandrolone-d3 (1.0 mg/ml in Acetonitrile)
Size:
1×1 ml