Lipomed Noroxycodone.HCl, 1 mg/ml in Methanol (as free base)
Catalog # 77976-110
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:52446-25-0
- Description:Noroxycodone.HCl, 1 mg/ml in Methanol (as free base)
- Size:1 ml