Carbohydrazide Activator Accessory Solution Pack
Product Details & Documents
The A-1800 Carbohydrazide Activator Accessory Solution Pack contains six 10 ml bottles of the activator solution necessary to perfrom analysis of carbohydrazide in water. This solution is found in K-1805 Carbohydrazide CHEMets®; Test Kit. This pack is meant as a refill for those who have already used their provided solution or who need to have solutions at multiple locations.
Specifications
Specifications
- Chemical method:PDTS
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)