TRC 7 alpha-Thiomethyl Spironolactone
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:38753-77-4
Specifications
Frequently Bought Together
Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Supplier: AMX INDUSTRIAL TRADE MX
Leakbuster™ Specimen Containers
Supplier: Starplex
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
Sodium chloride for biotechnology
Supplier: VWR
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Hydrogen peroxide 30% stabilized, CMOS for microelectronic use, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
QUICK-LOAD® Taq 2X Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
(R)-(-)-5-Oxotetrahydrofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Lysine Medium, BD Diagnostics
Supplier: BD
Lance Blades, Cincinnati Surgical
Supplier: CINCINNATI SURGICAL CO.
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade
Supplier: Thermo Scientific Chemicals