Dr. Ehrenstorfer Perfluorododecane
Catalog # 77976-061
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:307-59-5
- Description:Perfluorododecane
- Size:100 mg
- Shipping conditions:20 °C
- Formula:C₁₂F₂₆
- Formula weight:638,087 g/mol
- Storage temperature:20 °C
- Certification:ISO/IEC 17025