Order Entry
Canada
ContactUsLinkComponent
Paragon Scientific Polysulfide at 0.001% (10 ppm) in Light Mineral Oil
Paragon Scientific Polysulfide at 0.001% (10 ppm) in Light Mineral Oil
Catalog # 77975-850
Supplier:  LGC Standards
New Product
CAS Number:  
Paragon Scientific Polysulfide at 0.001% (10 ppm) in Light Mineral Oil
Catalog # 77975-850
Supplier:  LGC Standards
Supplier Number:  ALK-LMPS-020
New Product
CAS Number:  

Specifications

  • Description:
    Polysulfide at 0.001% (10 ppm) in light mineral oil
  • Size:
    100 ml
  • Storage temperature:
    RT

Specifications

About this item

  • Shipping conditions: Room temperature