Paragon Scientific IP 336, Sulfur set C in odorless kerosene: S @ 0.5, 1, 1.5, 2%
77975-809
New Product
:
- Matrix:Odourless Kerosene
- CAS No.:7704-34-9
- Description:IP 336, Sulfur set C in odorless kerosene: S @ 0.5, 1, 1.5, 2%
- Size:4×100 ml
- Certification:ISO 17034
- Storage conditions:RT
- Shipping conditions:RT
- Molecular weight:32.065