Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Mono(3-carboxypropyl) Phthalate-d4
TRC Mono(3-carboxypropyl) Phthalate-d4
TRC Mono(3-carboxypropyl) Phthalate-d4
Catalog # 77989-653
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC Mono(3-carboxypropyl) Phthalate-d4
Catalog # 77989-653
Supplier:
LGC Standards
Supplier Number:
TRC-M525637-2.5MG
New Product
CAS Number:
Specifications
CAS No.:
1346600-69-8
Description:
Mono(3-carboxypropyl) Phthalate-d4
Size:
2.5 mg
Specifications
About this item
Product Details
Product Details