TRC Hydroxy Dimetridazole
Supplier: LGC Standards
New Product
Certificates
About this item
Specifications
- CAS No.:936-05-0
Specifications
Frequently Bought Together
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Supplier: Thermo Scientific
Water, deionized VWR®
Supplier: VWR
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
Potassium dihydrogen phosphate ≥99% (by acidimetry), GR ACS, Supelco®
Supplier: MilliporeSigma
Hydrochloric acid 36,5 - 38,0% ACS
Supplier: Anachemia
Alconox® Powdered Precision Cleaners
Supplier: Alconox
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Air-Tite
Methanol, anhydrous ≥99.8% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
Water for HPLC, Supelco®
Supplier: MilliporeSigma
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Ethanol denatured, anhydrous
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
AccuStart™ II PCR Genotyping Kit, Quantabio
Supplier: Quantabio
Diisopropylamine ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Sol-M® Hypodermic Needles
Supplier: SOL-MILLENNIUM CANADA, INC. CA
Diethyl ether ≥98% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International