Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC (R)-N-(1-Carboxyethyl)-D-norvaline 1-Ethyl Ester
TRC (R)-N-(1-Carboxyethyl)-D-norvaline 1-Ethyl Ester
TRC (R)-N-(1-Carboxyethyl)-D-norvaline 1-Ethyl Ester
77979-472
:
LGC Standards
New Product
:
TRC (R)-N-(1-Carboxyethyl)-D-norvaline 1-Ethyl Ester
77979-472
:
LGC Standards
:
TRC-C178075-25MG
New Product
:
CAS No.:
145682-38-8
Description:
(R)-N-(1-Carboxyethyl)-D-norvaline 1-Ethyl Ester
Size:
25 mg