CHEMets® Ammonia/Monochloramine Refill (HBA)
77674-888
New Product
:
- Chemical method:Hydroxybenzyl Alcohol (HBA)
- Application:Drinking water;Clean surface water;Waste water;Storm water
The R-1402 Ammonia/Monochloramine CHEMets® refill (HBA) is a refill for K-1420 Ammonia CHEMets® test kit (HBA) or K-6802 Monochloramine CHEMets® test kit.It contains 30 self-filling CHEMets® ampoules packaged securely in cardboard trays and box. This product is meant to refill your CHEMets® if you already have the necessary comparator(s) and accessory solution(s).
- Method: Hydroxybenzyl Alcohol (HBA)