Dr. Ehrenstorfer Mass-Labelled PFC Injection Standards Solution 2 µg/ml in Methanol:Water
Catalog # 77976-055
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol:Water 96:4
- Description:Mass - Labelled PFC Injection standards solution 2 µg/ml in Methanol:Water
- Size:1.2 ml
- Shipping conditions:20 °C
- Storage temperature:−18 °C
- Certification:ISO 17034