Paragon Scientific Polysulfide at 0.3% (3000 ppm) in Light Mineral Oil
Catalog # 77975-861
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:Polysulfide at 0.3% (3000 ppm) in light mineral oil
- Size:100 ml
- Storage temperature:RT
Specifications
About this item
- Shipping conditions: Room temperature