$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
Standards
Organic Standards
TRC 21-Deacetoxy Deflazacort
TRC 21-Deacetoxy Deflazacort
77981-124
:
LGC Standards
New Product
:
TRC 21-Deacetoxy Deflazacort
77981-124
:
LGC Standards
:
TRC-D198000-10MG
New Product
:
CAS No.:
13649-88-2
Description:
21-Deacetoxy Deflazacort
Size:
10 mg