Dr. Ehrenstorfer Perfluoro-4-methoxybutanoic acid 13C2 (1,2-13C2) 50 µg/ml in Methanol:Water
Catalog # 77976-045
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol:Water 96:4
- Description:Perfluoro-4-methoxybutanoic acid 13C2 (1, 2 - 13C2) 50 µg/ml in Methanol:Water
- Size:1 ml
- Shipping conditions:20 °C
- Formula:¹³C₂C₃HF₉O₃
- Formula weight:282,031 g/mol
- Storage temperature:−18 °C
- Certification:ISO 17034